missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human FGFR2 [N549H], GST Tag Recombinant Protein

Artikelnummer. p-7181100
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
Pakningsstørrelse:
10µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 15900413

Brand: Invitrogen™ A33503

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This FGFR2 N549H recombinant human protein (285-end) was expressed in insect cells. FGFR2 N549H recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Cytoplasmic, Mutant The gene accession number for FGFR2 [N549H] is AAH39243.2.

FGFR2 receptor tyrosine kinase is a member of the fibroblast growth factor receptor family which plays a role in mitogenesis and differentiation. FGFR2 is a high-affinity receptor for acidic, basic and/or keratinocyte growth factor, and mutations in FGFR2 are associated with Crouzon syndrome, Pfeiffer syndrome, Craniosynostosis, Apert syndrome, Jackson-Weiss syndrome, Saethre-Chotzen syndrome, and syndromic craniosynostosis. FGFR2 is required for early postimplantation development between implantation and the formation of the egg cylinder. FGFR2 contributes to the outgrowth, differentiation, and maintenance of the inner cell mass.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P21802
Koncentration See Label
Til brug med (applikation) Kinase Assay
Formulering 50mM tris HCl with 25% glycerol, 0.25mM DTT, 0.1mM PMSF, 150mM NaCl, 10mM glutathione, 0.1mM EDTA and no preservative; pH 7.5
Gen-id (Entrez) 2263
Molekylvægt (g/mol) 74 kDa
Navn Human FGFR2 [N549H], GST Tag
pH-område 7.5
Oprensningsmetode Purified
Mængde 10 μg
Opbevaringskrav -80°C, Avoid Freeze/Thaw Cycles
Regulatorisk status RUO
Gene Alias FGFR2
Fælles navn FGFR2 (N549H Mutant)
Gen symbol FGFR2
KinaseFamily FGF Receptors
Protein længde 285-end
Produkttype Protein
Konjugeret Unconjugated
Arter Human
Rekombinant Recombinant
Protein tag GST-tag
Sekvens 285-end
Indhold og opbevaring -80°C, Avoid Freeze/Thaw Cycles
Forsendelsestilstand Dry Ice
Udtrykssystem Insect cells
Protein familie Kinases & Inhibitors
Protein form Recombinant, Active, Cytoplasmic, Mutant
Form Liquid
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.