Få mere at vide
Invitrogen™ Human FGFR2 [N549H], GST Tag Recombinant Protein
Beskrivelse
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This FGFR2 N549H recombinant human protein (285-end) was expressed in insect cells. FGFR2 N549H recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Cytoplasmic, Mutant The gene accession number for FGFR2 [N549H] is AAH39243.2.
Tekniske data
Tekniske data
| Adgangsnummer | P21802 |
| Koncentration | See Label |
| Til brug med (applikation) | Kinase Assay |
| Formulering | 50mM tris HCl with 25% glycerol, 0.25mM DTT, 0.1mM PMSF, 150mM NaCl, 10mM glutathione, 0.1mM EDTA and no preservative; pH 7.5 |
| Gen-id (Entrez) | 2263 |
| Molekylvægt (g/mol) | 74 kDa |
| Navn | Human FGFR2 [N549H], GST Tag |
| pH-område | 7.5 |
| Oprensningsmetode | Purified |
| Mængde | 10 μg |
| Vis mere |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.