missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13254-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the fibroblast growth factor binding protein family. The encoded protein is a serum protein that is selectively secreted by cytotoxic lymphocytes and may be involved in cytotoxic lymphocyte-mediated immunity. An increase in the amount of gene product may be associated with atopic asthma and mild extrinsic asthma.Specifications
FGFBP2 | |
Polyclonal | |
Unconjugated | |
FGFBP2 | |
FGF binding protein 2, FGF BP2, FGFBP 2, FGFBP2, HBp17 related protein, HBp17 RP, HBP17RP, KSP37 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83888 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BYJ0 | |
FGFBP2 | |
FGFBP2 Fusion Protein Ag4036 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FGFBP2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction