Få mere at vide
Novus Biologicals™ FGF basic/FGF2/bFGF Antibody (HuGAL-F2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28540-50ug
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| FGF basic/FGF2/bFGF | |
| Monoclonal | |
| Unconjugated | |
| P09038 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| HuGAL-F2 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| FGF2 | |
| 50 μg | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 2247 | |
| Human | |
| Purified |