Learn More
FFP2 Disposable Respirator Face Mask
FFP2 Disposable respirator to protect against inhalation of droplets and particles. Available with head band and without exhalation valve.
Brand:  Roesner Mautby Meditrade ZH3161
Additional Details : Weight : 0.95000kg
Product Code. 17176314
  • 15.00 DKK / Pack of 40
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
Alert:
Compliance
This product complies with the essential requirements for health and safety as they are provided by the European Regulation (EU) 2016/425 for Personal Protective Equipment.
Description
Description
  • Tested and certified FFP2 disposable respirator
  • Without exhalation valve
  • Head bands enable a more secure, tighter and comfortable fit
  • Head bands make wearing glasses more comfortable and enable good compatibility with various eye protection products
Specifications
Specifications
CE Approved, EN 149:2001+A1:2009 FFP2 NR
Single-use
Face Piece: Polypropylene and Polyester; Head Band: Polyurethane and Polyester; Nosepiece: Metal
One Size
Non-sterile
Flat-fold
FFP2 NR
Particulate Respirator with Double Head Bands