missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FFN 270

Description
FFN 270 is a fluorescent false neurotransmitter (FFN). Fluorescent substrate for NET and VMAT2. Labels noradrenergic neurons and their synaptic vesicles, and enables imaging of synaptic vesicle content release from specific axonal sites in vivo. Selectively labels NA neurons over other monoamine and CNS targets. Exhibits two resolved absorption/excitation maxima depending on solvent pH (excitation maxima: 320 nm or 365 nm, emission maxima: 475 nm).
Specifications
Specifications
| Kemisk navn eller materiale | 3-(2-Aminoethyl)-8-fluoro-7-hydroxy-2H-chromen-2-one hydrochloride |
| CAS | 2341841-05-0 |
| Mængde | 10 mg |
| Opløselighed | Soluble to 100mM in DMSO |
| Mål | Fluorescent Transporter Probes |
| Molekylær formel | C11H10FNO3.HCl |
| Formel vægt | 259.66 |
| Fysisk form | Solid |
| Farve | Beige |
| Produkttype | Fluorescent substrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?