Få mere at vide
Tocris Bioscience™ FFN 102 mesylate
GREENER_CHOICE
Selective fluorescent substrate of DAT and VMAT2
Brand:  Tocris Bioscience™ 5200/10
Encompass
Produktkode 15776645
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
FFN 102 mesylate is a pH responsive fluorescent false neurotransmitter (FFN). Selective dopamine transporter (DAT) and VMAT2 substrate. Exhibits no significant binding to a panel of 38 CNS receptors, including dopamine and serotonin receptors. Inhibits dopamine uptake. Excitation maxima are 340 nm at pH 5 and 370 nm at pH 7.5. Emission maximum is 435 nm at both pH 5 and 7.5.
Tekniske data
Tekniske data
>99%
1883548-92-2
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only