missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ FEZ2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17770514
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17770514 200 μL
17743394 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17770514 Leverandør Invitrogen™ Leverandørnr. PA5119105

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

FEZ2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Fasciculation and elongation protein zeta-2 (FEZ2) is a homolog to the mammalian FEZ1, itself an ortholog of the C. elegans UNC-76. In contrast to FEZ1, FEZ2 mRNA is widely expressed in mouse tissues. FEZ2 interacts with protein kinase C (PKC)-zeta substrate and induces neurite extension of PC12 cells when co-expressed with a constitutively active form of PKC-zeta, suggesting FEZ2 may play an important role in the morphological changes of various cells by associating with PKC-zeta in a tissue non-specific manner. FEZ2 can interact with FEZ1 through its c-terminal regions and especially its coiled-coil region. At least two isoforms of FEZ2 are known to exist.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen FEZ2
Applikationer Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene FEZ2
Genadgangsnr. P97578, Q9UHY8
Gene Alias 9030616F16; D17Ertd315e; fasciculation and elongation protein zeta 2; fasciculation and elongation protein zeta 2 (zygin II); fasciculation and elongation protein zeta-2; FEZ2; HUM3CL; pre-T/NK cell associated protein (3Cl); synaptotagmin interacting protein zyginII; Zrp; zygin 2; zygin II; Zygin-2; zygin-related protein types I/II
Gensymboler FEZ2
Værtsarter Rabbit
Immunogen Recombinant protein Fasciculation And Elongation Protein Zeta 2. The antigen corresponds to amino acid range 1-375 of the target protein.
Oprensningsmetode Antigen affinity chromatography, Protein A
Mængde 200 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 94269, 9637
Målarter Human, Rat
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.