missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin B Antibody (110), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90643UV
Additional Details : Weight : 0.00970kg
Description
Fetuin B Monoclonal antibody specifically detects Fetuin B in Mouse samples. It is validated for ELISA.Specifications
Fetuin B | |
Monoclonal | |
DyLight 350 | |
FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fetuin B (Q9QXC1-1; Met 1-Pro 388). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human FETUB | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
110 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
26998 | |
Mouse | |
Purified |