missing translation for 'onlineSavingsMsg'
Få mere at vide

Ferroprotoporphyrin IX, TRC

Artikelnummer. 30137591
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137591 25 mg
30133896 50 mg
30005205 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30137591 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCF30761025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14875-96-8
Molekylær formel C34H32FeN4O42
Molekylvægt (g/mol) 616.49
Synonym 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic Acid Ferrous Complex, Ferroheme, Hem Fe, Heme, Heme B, Hydrogen [7,12-Diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropanoato(4-)-κN21,κN22,κN23,κN24]ferrate (heme), Iron Protoporphyrin, Iron Protoporphyrin IX, Iron(II) Protoporphyrin IX, NSC 267100, Protoheme, Protoheme IX, Reduced Hematin, [Dihydrogen 3,7,12,17-tetramethyl-8,13-divinyl-2,18-porphinedipropionato(2-)]iron, (SP-4-2)-[7,12-diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropanoato(4-)-N21,N22,N23,N24]-Ferrate(2-) Dihydrogen, Protoporphyrin Iron Complex, 7,12-diethenyl-3,8,13,17-tetramethyl-21H,23H-Porphine-2,18-dipropanoic Acid Iron Complex, (SP-4-2)-[7,12-diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropanoato(4-)-κN21,κN22,κN23,κN24]-ferrate(2-) Dihydrogen;
SMIL OC(CCC1=C(C=C2[N]3=C4C(C)=C2CCC(O)=O)[N]([Fe]35[N]6=C7C=C(C(C=C)=C8C)[N]5C8=C4)C(C=C6C(C=C)=C7C)=C1C)=O

Specifications

Kemisk navn eller materiale Ferroprotoporphyrin IX
Mængde 25 mg
InChI formel InChI=1S/C34H34N4O4.Fe/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);/q;+2/p-2/b25-13?,26-13-,27-14?,28-15-,29-14+,30-15?,31-16-,32-16?;
Formel vægt 616.1773
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.