Få mere at vide
Ferrocene-d10, CDN
Artikelnummer. 30327867
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327867 0.25 g
30316348 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327867 Leverandør CDN Leverandørnr. CDND11630.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ferrocene-d10
CAS 12082-87-0
Molekylær formel 2 C5 D5 . Fe
Mængde 0.25 g
Synonym Ferrocene-1,1',2,2',3,3',4,4',5,5'-d10 (ACI), Ferrocene-d10 (8 CI, 9 CI), Iron, di(cyclopentadienyl-d5)- (6 CI, 7 CI), Decadeuteroferrocene, Ferrocene-D10
InChI formel InChI=1 S/2C5H5.Fe/c2*1-2-4-5-3-1;/h2*1-5 H;/i2*1 D,2 D,3 D,4 D,5 D;
SMIL [Fe].[2 H][C]1 [C]([2 H])[C]([2 H])[C]([2 H])[C]1[2 H].[2 H][C]2 [C]([2 H])[C]([2 H])[C]([2 H])[C]2[2 H]
IUPAC navn iron;1,2,3,4,5-pentadeuteriocyclopentane
Molekylvægt (g/mol) 196.093
Formel vægt 196.076 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity