missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ferritin Light Chain Antibody (FTL/1386), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54574V
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Ferritin Light Chain Monoclonal antibody specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Tekniske data
| Ferritin Light Chain | |
| Monoclonal | |
| DyLight 405 | |
| Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3 | |
| Recombinant fragment (around aa 38-165) of human Ferritin Light Chain protein (exact sequence is proprietary) (Uniprot: P02792) | |
| 100 μg | |
| Cancer | |
| 2512 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| FTL/1386 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion