Learn More
Thermo Scientific™ XceI (NspI) (10 U/μL)
Cut at RCATG^Y sites with XceI (NspI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: BstNSI, NspI).
810.00 DKK - 3260.00 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10387799
|
Thermo Scientific™
ER1471 |
500 U |
810.00 DKK
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10642001
|
Thermo Scientific™
ER1472 |
2500 U |
3260.00 DKK
2500 units |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Lambda DNA digested with XceI (NspI), 0.7% agarose, 32 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
- Superior quality -stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities
Specifications
10 U/μL |
For Research Use Only. Not for use in diagnostic procedures.