Learn More
Thermo Scientific™ Rapid DNA Ligation Kit Product Code.: 10775841
Thermo Scientific™ Rapid DNA Ligation Kit
Product Code. p-4530525 Shop All Thermo Scientific Products
50 Reactions, 50 reactions
Click to view available options
Quantity:
150 Reactions
50 Reactions
Unit Size:
150 reactions
50 reactions
Product Code. 10775841
Brand: Thermo Scientific™ K1422
Edge
Enable fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature.
The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
The kit contains T4 DNA ligase and a specially formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
  • Fast—ligation of either sticky- or blunt-end DNA in only 5 minutes
  • Convenient—reaction mixture can be used directly for bacterial transformation
Recommended for:
Routine cloning experiments; Blunt-end cloning; Library construction; TA cloning.
Note:
Lyophilized bacterial strains GM2163 (†M0099) and JM107 (†M0109) can be provided free of charge upon request, with any purchase. Prior to electroporation it is necessary to inactivate T4 DNA ligase (1) by chloroform extraction.
TRUSTED_SUSTAINABILITY
Specifications
Buffer 5X Rapid Ligation Buffer
Enzyme DNA Ligase
Product Type Rapid DNA Ligation Kit
Quantity 50 Reactions
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2822089 Certificate TypeCertificate of Analysis Date29/01/2025 Product CodeK1422
Lot Number2825198 Certificate TypeCertificate of Analysis Date29/01/2025 Product CodeK1422
Lot Number2792572 Certificate TypeCertificate of Analysis Date29/01/2025 Product CodeK1422
Lot Number2841211 Certificate TypeCertificate of Analysis Date29/01/2025 Product CodeK1422
Lot Number2866370 Certificate TypeCertificate of Analysis Date28/01/2025 Product CodeK1422
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ Rapid DNA Ligation Kit >