Learn More
Thermo Scientific™ NoLimits 600 bp DNA Fragment
GREENER_CHOICE
NoLimits is a collection of 36 chromatography-purified individual DNA fragments ranging from 10 to 20,000 bp for applications where a custom DNA size standard is required.
Brand:  Thermo Scientific™ SM1461
493.35 DKK valid until 2025-07-04
Use promo code "24112" to get your promotional price.
Product Code. 10170780
  • 740.00 DKK / 10µg
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
Thermo Scientific NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology. Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agarose
No
Agarose Gels
NoLimits
DNA Fragment
10 to 20000 bp
Sustainable packaging
10 μg
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2830026 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM1461
Lot Number2790640 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM1461
Lot Number2894128 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSM1461
Lot Number2901882 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSM1461
Lot Number2907819 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSM1461
Promotions
Promotions
  • Multiple Manufacturers
    Exclusive savings, limited-time offer.
    Promo Code: 24112
    Expires2025-07-04
    New Offer
    Save Now
Product Content Correction
Product Title
Thermo Scientific™ NoLimits 600 bp DNA Fragment > 10μg