missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bpu1102I (BlpI) (10 U/μL)

Cut at GC^TNAGC sites with Bpu1102I (BlpI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: BlpI, Bsp1720I, CelII).

Brand:  Thermo Scientific™ ER0092

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10450460

  • 1348.00 DKK / 1000 units
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G C▵T N A G C...3'

3'...C G A N T▵C G...5'

Conditions for 100% Activity:

  • 1X Buffer Tango™: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Bpu1102I is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
  •  

Ligation and Recleavage:

  • After 50-fold overdigestion with Bpu1102I, more than 80% of the DNA fragments can be ligated in a reaction mixture containing 20-40 u of T4 DNA Ligase/1μg of fragments and 10% PEG. More than 95% of these can be recut

Methylation Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — no effect.
  • EcoKI: never overlaps — no effect.
  • EcoBI: may overlap — effect not determined

Digestion of Agarose-embedded DNA:

  • Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours
  • Compatible Ends: Bpu10I, Eco81I, HpyF3I
Specifications

Specifications

10 U/μL
Bpu1102I (BlpI)
Yes
No
Restriction Enzyme
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X Buffer Tango
37°C
1000 U
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.