Få mere at vide
Feprazone, LoGiCal
Artikelnummer. 30460808
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460808 10 mg
1 options
Artikelnummer. 30460808 Leverandør LoGiCal Leverandørnr. LGCFOR0691.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Feprazone
CAS 30748-29-9
Analyt- eller komponentnavne Feprazone
Forsendelsestilstand Room Temperature
Molekylær formel C20 H20 N2 O2
Certificeringer/compliance ISO 9001
Synonym 3,5-Pyrazolidinedione, 4-(3-methyl-2-buten-1-yl)-1,2-diphenyl-, 3,5-Pyrazolidinedione, 4-(3-methyl-2-butenyl)-1,2-diphenyl- (8CI,9CI), 4-(2-Isopentenyl)-1,2-diphenyl-3,5-pyrazolidinedione, 4-(3-Methyl-2-butenyl)-1,2-diphenyl-3,5-pyrazolidinedione, 4-Prenyl-1,2-diphenyl-3,5-dioxopyrazolidine, 4-Prenyl-1,2-diphenyl-3,5-pyrazolidinedione, Analud, DA 2370, Fenilprenazone, Feprazone, Methrazone, Metrazone, Phenylprenazone, Pheprazone, Prenazone, Zepelin
InChI formel InChI=1S/C20H20N2O2/c1-15(2)13-14-18-19(23)21(16-9-5-3-6-10-16)22(20(18)24)17-11-7-4-8-12-17/h3-13,18H,14H2,1-2H3
Løsningstype Neat
SMIL CC(=CCC1C(=O)N(N(C1=O)c2ccccc2)c3ccccc3)C
IUPAC navn 4-(3-methylbut-2-enyl)-1,2-diphenylpyrazolidine-3,5-dione
Industry Type Clinical Testing
Molekylvægt (g/mol) 320.38
Mængde 10 mg
Formel vægt 320.1525 g/mol