Learn More
Fenthion, TRC
Product Code. 30134386
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134386 25 g 25g
30134513 10 g 10g
17927979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30134386 Supplier Toronto Research Chemicals Supplier No. TRCF27800025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55-38-9
Molecular Formula C10 H15 O3 P S2
Molecular Weight (g/mol) 278.33
Synonym Phosphorothioic acid, O,O-dimethyl O-[3-methyl-4-(methylthio)phenyl] ester (9CI, ACI), m-Cresol, 4-(methylthio)-, O-ester with O,O-di-Me phosphorothioate (6CI), Phosphorothioic acid (H3PO3S), O,O-dimethyl O-4-(methylthio)-m-tolyl ester (6CI), Phosphorothioic acid, dimethyl [4-(methylthio)-m-tolyl] ester (7CI), Phosphorothioic acid, O,O-dimethyl O-[4-(methylthio)-m-tolyl] ester (8CI), B 29493, BAY 29493, Baycid, Bayer 9007, Baytex, Dimethyl 4-methylthio-m-tolyl phosphorothioate, ENT 25540, Entex, Fenthion, Fenthion-methyl, Lebaycid, Mercaptofos, Mercaptophos, MeSH ID: D005284, MPP, MPP (pesticide), O,O-Dimethyl O-(3-methyl-4-methylmercaptophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-methylthiophenyl) thiophosphate, O,O-Dimethyl O-(4-methylthio-3-methylphenyl) phosphorothioate, O,O-Dimethyl O-4-(methylmercapto)-3-methylphenyl phosphorothioate, O,O-Dimethyl O-4-(methylmercapto)-3-methylphenyl thiophosphate, O,O-Dimethyl O-[3-methyl-4-(methylthio)phenyl] phosphorothioate, O,O-Dimethyl O-[4-(methylthio)-m-tolyl] phosphorothioate, O,O-Dimethyl O-[4-(methylthio)-m-tolyl] thiophosphate, OMS 2, Phenthion, Queletox, S 1752, Spotton, Sulfidophos, Talodex, Tiguvon
IUPAC Name dimethoxy-(3-methyl-4-methylsulfanylphenoxy)-sulfanylidene-λ^{5}-phosphane
SMILES COP(=S)(OC)Oc1ccc(SC)c(C)c1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C10H15O3PS2/c1-8-7-9(5-6-10(8)16-4)13-14(15,11-2)12-3/h5-7H,1-4H3
Formula Weight 278.02
Chemical Name or Material Fenthion