Få mere at vide
Fenthion-d6 (O,O-dimethyl-d6), CDN
Artikelnummer. 30319729
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30319729 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319729 Leverandør CDN Leverandørnr. CDND64620.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fenthion-d6
CAS 1189662-83-6
Molekylær formel C10 2H6 H9 O3 P S2
Mængde 0.01 g
InChI formel InChI=1 S/C10H15O3PS2/c1-8-7-9(5-6-10(8)16-4)13-14(15,11-2)12-3/h5-7 H,1-4H3/i2D3,3D3
SMIL [2 H]C([2 H])([2 H])OP(=S)(Oc1ccc(SC)c(C)c1)OC([2 H])([2 H])[2 H]
IUPAC navn (3-methyl-4-methylsulfanylphenoxy)-sulfanylidene-bis(trideuteriomethoxy)-λ^{5}-phosphane
Molekylvægt (g/mol) 284.36
Formel vægt 284.0577 g/mol
Procent renhed 99 atom % D, min 95% Chemical Purity