Få mere at vide
Fensulfothion-sulfone D10 (diethyl D10), Dr. Ehrenstorfer
Artikelnummer. 30316834
Skift visning
Mængde:
10 mg
Pakningsstørrelse:
10mg
Artikelnummer. Mængde unitSize
30316834 10 mg 10mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316834 Leverandør Dr. Ehrenstorfer Leverandørnr. DREC13570025
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fensulfothion Sulfone-d10
CAS 2732982-58-8
Molekylær formel C11H7D10O5PS2
Mængde 10 mg
Synonym O,O-Diethyl O-[4-(Methylsulfonyl)phenyl] Ester Phosphorothioic Acid-d10, Dasanit Sulfone-d10, Dasanit Sulphone-d10, O,O-Diethyl O-(4-Methylsulfonylphenyl) Phosphorothioate-d10, Fensulfothion-sulfone diethyl-d10, Fensulfothion-sulfone-D10 (diethyl-D10)
InChI formel InChI=1S/C11H17O5PS2/c1-4-14-17(18,15-5-2)16-10-6-8-11(9-7-10)19(3,12)13/h6-9H,4-5H2,1-3H3/i1D3,2D3,4D2,5D2
SMIL O=S(C1=CC=C(OP(OC([2H])(C([2H])([2H])[2H])[2H])(OC([2H])(C([2H])([2H])[2H])[2H])=S)C=C1)(C)=O
IUPAC navn O,O-bis(ethyl-d5) O-(4-(methylsulfonyl)phenyl) phosphorothioate
Molekylvægt (g/mol) 334.42
Formel vægt 334.09 g/mol