Få mere at vide
Fensulfothion Sulfid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fensulfothion-sulfide |
| CAS | 3070-15-3 |
| Molekylær formel | C11 H17 O3 P S2 |
| Mængde | 100 mg |
| Synonym | Phosphorothioic acid, O,O-diethyl O-[4-(methylthio)phenyl] ester, Phosphorothioic acid, O,O-diethyl O-[p-(methylthio)phenyl] ester (6CI,7CI,8CI), Phenol, p-(methylthio)-, O-ester with O,O-diethyl phosphorothioate (8CI), Fensulfothion sulfide, O,O-Diethyl O-[p-(methylthio)phenyl] phosphorothioate |
| InChI formel | InChI=1S/C11H17O3PS2/c1-4-12-15(16,13-5-2)14-10-6-8-11(17-3)9-7-10/h6-9H,4-5H2,1-3H3 |
| SMIL | CCOP(=S)(OCC)Oc1ccc(SC)cc1 |
| IUPAC navn | diethoxy-(4-methylsulfanylphenoxy)-sulfanyliden-λ^{5}-fosfan |
| Molekylvægt (g/mol) | 292.35 |
| Formel vægt | 292.0357 |