Learn More
Fensulfothion, Honeywell Fluka™
Product Code. 16694534 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
16694534 100 mg 100mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 16694534 Supplier Honeywell Fluka Supplier No. R1741100MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fensulfothion
CAS 115-90-2
CAS Max % ≤ 100%
Density 1.202 g/cm3 (20°C)
Beilstein 2219515
Color Light Yellow
Melting Point <25°C
Boiling Point 138°C to 141°C (0.01 hPa)
Synonym Diethyl 4-(methylthio)phenyl phosphate
Molecular Weight (g/mol) 308.35 g/mol
Quantity 100 mg