Få mere at vide
Fenpropimorph-d3, TRC
Artikelnummer. 30407936
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30407936 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407936 Leverandør Toronto Research Chemicals Leverandørnr. TRCF24950210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fenpropimorph-d3
CAS 1292815-71-4
Renhedsgrad noter HPLC
Molekylær formel C20H30D3NO
Mængde 10 mg
Synonym (2R,6S)-rel-4-[3-[4-(1,1-Dimethylethyl)phenyl]-2-(methyl-d3)propyl]-2,6-dimethylmorpholine, cis-4-[3-[4-(1,1-Dimethylethyl)phenyl]-2-(methyl-d3)propyl]-2,6-dimethyl- morpholine, cis-4-[3-(4-tert-Butylphenyl)-2-(methyl-d3)propyl]-2,6-dimethyl- morpholine, BAS 42100F-d3, Corbel-d3, Forbel 750-d3, Mistral-d3
InChI formel InChI=1S/C20H33NO/c1-15(12-21-13-16(2)22-17(3)14-21)11-18-7-9-19(10-8-18)20(4,5)6/h7-10,15-17H,11-14H2,1-6H3/t15?,16-,17+/i1D3
SMIL CC(C)(C)C(C=C1)=CC=C1CC(C([2H])([2H])[2H])CN2C[C@@H](C)O[C@@H](C)C2
Molekylvægt (g/mol) 306.5
Formel vægt 306.275
Procent renhed >95