Få mere at vide
Fenpiprane Hydrochloride, LoGiCal
Artikelnummer. 30461948
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461948 10 mg
1 options
Artikelnummer. 30461948 Leverandør LoGiCal Leverandørnr. LGCFOR3073.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fenpiprane Hydrochloride
CAS 3329-14-4
Analyt- eller komponentnavne Fenpiprane Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C20 H25 N . Cl H
Certificeringer/compliance ISO 9001
Synonym Piperidine, 1-(3,3-diphenylpropyl)-, hydrochloride (1:1), Piperidine, 1-(3,3-diphenylpropyl)-, hydrochloride (6CI,7CI,8CI,9CI), 1,1-Diphenyl-3-piperidinopropane hydrochloride, 1-(3,3-Diphenylpropyl)piperidine hydrochloride, Fenpiprane hydrochloride, N-(3,3-Diphenylpropyl)piperidine hydrochloride
InChI formel InChI=1S/C20H25N.ClH/c1-4-10-18(11-5-1)20(19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;/h1-2,4-7,10-13,20H,3,8-9,14-17H2;1H
Løsningstype Neat
SMIL Cl.C(CN1CCCCC1)C(c2ccccc2)c3ccccc3
IUPAC navn 1-(3,3-diphenylpropyl)piperidine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 315.88
Mængde 10 mg
Formel vægt 315.1754 g/mol