Få mere at vide
Fenmetramide, LoGiCal
Artikelnummer. 30462126
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462126 10 mg
1 options
Artikelnummer. 30462126 Leverandør LoGiCal Leverandørnr. LGCFOR3348.00
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Fenmetramide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fenmetramide
CAS 5588-29-4
Analyt- eller komponentnavne Fenmetramide
Forsendelsestilstand Room Temperature
Molekylær formel C11 H13 N O2
Certificeringer/compliance ISO 9001
Synonym 3-Morpholinone, 5-methyl-6-phenyl-, 5-Methyl-3-oxo-6-phenyl-3-morpholin, 5-Methyl-6-phenyl-3-morpholinone, Feninetramide, Fenmetramide, McN 1075, NSC 169876
InChI formel InChI=1 S/C11H13NO2/c1-8-11(14-7-10(13)12-8)9-5-3-2-4-6-9/h2-6,8,11 H,7H2,1H3,(H,12,13)
SMIL CC1NC(=O)COC1c2ccccc2
IUPAC navn 5-methyl-6-phenylmorpholin-3-one
Industry Type Forensics
Molekylvægt (g/mol) 191.23
Mængde 10 mg
Formel vægt 191.0946 g/mol