missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Fenitrothion, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fenitrothion |
| CAS | 122-14-5 |
| Molekylær formel | C9 H12 N O5 P S |
| Mængde | 1 g |
| Synonym | Phosphorothioic acid, O,O-dimethyl O-(4-nitro-m-tolyl) ester (6CI,8CI), 8057HC, Accothion, Agria 1050, Agriya 1050, Agrothion, Arbogal, BAY 41831, Bayer 41831, Bayer S 5660, CL 47300, CP 47114, Dimethyl 4-nitro-m-tolyl phosphorothionate, EI 47300, ENT 25,715, Fenition, Fenitrothion, Folithion, Folithion EC 50, Insectigas F, MEP, MEP (pesticide), Metathion, Metathion E 50, Metathione, Metathionine E 50, Metation, Metation E 50, Methadion, Methylnitrophos, Mglawik F, Monsanto CP 47114, Nitrophos, Nuvanol, O,O-DiMe O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(4-nitro-3-methylphenyl)thiophosphate, O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate, O,O-Dimethyl O-4-nitro-m-tolyl thiophosphate, OMS 43, Oleometathion, Oleosumifene, Ovadofos, Owadofos, Owadophos, Paluthion, Phenitrothion, S 112A, S 5660, ShaMingSong, Sumi oil, Sumifene, Sumigran, Sumithion, Sumithion 20F, Sumithion 20MC, Sumithion 50EC, Sumithion Super, Sumithion Super 1000EC, Super Sumithion, Tionfos 50 LE, Verthion |
| InChI formel | InChI=1S/C9H12NO5PS/c1-7-6-8(4-5-9(7)10(11)12)15-16(17,13-2)14-3/h4-6H,1-3H3 |
| SMIL | COP(=S)(OC)Oc1ccc(c(C)c1)[N+](=O)[O-] |
| IUPAC navn | dimethoxy-(3-methyl-4-nitrophenoxy)-sulfanylidene-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 277.23 |
| Formel vægt | 277.0174 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?