missing translation for 'onlineSavingsMsg'
Få mere at vide

Fenitrothion, TRC

Artikelnummer. 30403442
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30403442 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30403442 Leverandør Toronto Research Chemicals Leverandørnr. TRCF247850250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Fenitrothion
CAS 122-14-5
Molekylær formel C9 H12 N O5 P S
Mængde 250 mg
Synonym Phosphorothioic acid, O,O-dimethyl O-(4-nitro-m-tolyl) ester (6CI,8CI), 8057HC, Accothion, Agria 1050, Agriya 1050, Agrothion, Arbogal, BAY 41831, Bayer 41831, Bayer S 5660, CL 47300, CP 47114, Dimethyl 4-nitro-m-tolyl phosphorothionate, EI 47300, ENT 25,715, Fenition, Fenitrothion, Folithion, Folithion EC 50, Insectigas F, MEP, MEP (pesticide), Metathion, Metathion E 50, Metathione, Metathionine E 50, Metation, Metation E 50, Methadion, Methylnitrophos, Mglawik F, Monsanto CP 47114, Nitrophos, Nuvanol, O,O-DiMe O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(4-nitro-3-methylphenyl)thiophosphate, O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate, O,O-Dimethyl O-4-nitro-m-tolyl thiophosphate, OMS 43, Oleometathion, Oleosumifene, Ovadofos, Owadofos, Owadophos, Paluthion, Phenitrothion, S 112A, S 5660, ShaMingSong, Sumi oil, Sumifene, Sumigran, Sumithion, Sumithion 20F, Sumithion 20MC, Sumithion 50EC, Sumithion Super, Sumithion Super 1000EC, Super Sumithion, Tionfos 50 LE, Verthion
InChI formel InChI=1S/C9H12NO5PS/c1-7-6-8(4-5-9(7)10(11)12)15-16(17,13-2)14-3/h4-6H,1-3H3
SMIL COP(=S)(OC)Oc1ccc(c(C)c1)[N+](=O)[O-]
IUPAC navn dimethoxy-(3-methyl-4-nitrophenoxy)-sulfanylidene-λ^{5}-phosphane
Molekylvægt (g/mol) 277.23
Formel vægt 277.0174
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.