Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fenitrothion |
| CAS | 122-14-5 |
| CAS Max % | 1 |
| Tæthed | 1.320 g/cm3 (20°C) |
| Flammepunkt | 157°C |
| Beilstein | 8983553 |
| Farve | Yellow |
| Smeltepunkt | 0.3°C |
| Kogepunkt | 140°C to 145°C |
| Synonym | O,O-dimethyl O-4-nitro-m-tolyl phosphorothioate |
| Vis mere |