missing translation for 'onlineSavingsMsg'
Få mere at vide

Fenitrothion 100 μg/mL in Cyclohexane, Dr. Ehrenstorfer

Artikelnummer. 30310991
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 ml
Pakningsstørrelse:
1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310991 1 ml 1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30310991 Leverandør Dr. Ehrenstorfer Leverandørnr. DREXA13480000CY

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Fenitrothion
CAS 122-14-5
Analyt- eller komponentnavne Fenitrothion
Forsendelsestilstand Room Temperature
Molekylær formel C9 H12 N O5 P S
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
Synonym Phosphorothioic acid, O,O-dimethyl O-(4-nitro-m-tolyl) ester (6CI,8CI), 8057HC, Accothion, Agria 1050, Agriya 1050, Agrothion, Arbogal, BAY 41831, Bayer 41831, Bayer S 5660, CL 47300, CP 47114, Dimethyl 4-nitro-m-tolyl phosphorothionate, EI 47300, ENT 25,715, Fenition, Fenitrothion, Folithion, Folithion EC 50, Insectigas F, MEP, MEP (pesticide), Metathion, Metathion E 50, Metathione, Metathionine E 50, Metation, Metation E 50, Methadion, Methylnitrophos, Mglawik F, Monsanto CP 47114, Nitrophos, Nuvanol, O,O-DiMe O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(4-nitro-3-methylphenyl)thiophosphate, O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate, O,O-Dimethyl O-4-nitro-m-tolyl thiophosphate, OMS 43, Oleometathion, Oleosumifene, Ovadofos, Owadofos, Owadophos, Paluthion, Phenitrothion, S 112A, S 5660, ShaMingSong, Sumi oil, Sumifene, Sumigran, Sumithion, Sumithion 20F, Sumithion 20MC, Sumithion 50EC, Sumithion Super, Sumithion Super 1000EC, Super Sumithion, Tionfos 50 LE, Verthion
InChI formel InChI=1S/C9H12NO5PS/c1-7-6-8(4-5-9(7)10(11)12)15-16(17,13-2)14-3/h4-6H,1-3H3
Løsningstype Single Solution
SMIL COP(=S)(OC)Oc1ccc(c(C)c1)[N+](=O)[O-]
IUPAC navn dimethoxy-(3-methyl-4-nitrophenoxy)-sulfanylidene-λ^{5}-phosphane
Opløsningsmiddel eller Matrix Cyclohexane
Industry Type Environmental Testing
Molekylvægt (g/mol) 277.23
Mængde 1 ml
Formel vægt 277.0174 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.