Få mere at vide
Fenitrooxon-d6 (O,O-dimethyl-d6), CDN
Artikelnummer. 30316757
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30316757 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316757 Leverandør CDN Leverandørnr. CDND64180.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fenitrooxon-d6
CAS 1185155-54-7
Molekylær formel C9 2H6 H6 N O6 P
Mængde 0.01 g
Synonym Fenitrothion-oxon D6 (O,O-dimethyl D6), Fenitrooxon-d6
InChI formel InChI=1 S/C9H12NO6P/c1-7-6-8(4-5-9(7)10(11)12)16-17(13,14-2)15-3/h4-6 H,1-3H3/i2D3,3D3
SMIL [2 H]C([2 H])([2 H])OP(=O)(Oc1ccc(c(C)c1)[N+](=O)[O-])OC([2 H])([2 H])[2 H]
IUPAC navn (3-methyl-4-nitrophenyl) bis(trideuteriomethyl) phosphate
Molekylvægt (g/mol) 267.21
Formel vægt 267.0779 g/mol
Procent renhed 99 atom % D, min 95% Chemical Purity