Få mere at vide
Fenirofibrate-d6, TRC
Artikelnummer. 30406747
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30406747 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406747 Leverandør Toronto Research Chemicals Leverandørnr. TRCF2477021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fenirofibrate-d6
CAS 1189423-29-7
Molekylær formel C17H11D6ClO4
Mængde 1 mg
Synonym 2-[4-[(4-Chlorophenyl)hydroxymethyl]phenoxy]-2-methyl-propanoic Acid-d6, Hydroxymethyl Fenofibric Acid-d6, LF 2151, LF 433-d6
InChI formel InChI=1S/C17H17ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10,15,19H,1-2H3,(H,20,21)/i1D3,2D3
SMIL OC(C1=CC=C(OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C(O)=O)C=C1)C2=CC=C(Cl)C=C2
Molekylvægt (g/mol) 326.8