Få mere at vide
Fenfluramine HCl, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fenfluramine Hydrochloride |
| CAS | 404-82-0 |
| Analyt- eller komponentnavne | Fenfluramine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H16 F3 N . Cl H |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | 1-(m-Trifluoromethylphenyl)-2-(ethylamino)propane hydrochloride, Fenfluramine hydrochloride, Ganal, HHR-965, Ponderal, Ponderax, Ponderex, Pondimin, Pondmin, Racemic fenfluramine hydrochloride, S 768, dl-Fenfluramine hydrochloride, Benzeneethanamine, N-ethyl-alpha-methyl-3-(trifluoromethyl)-, hydrochloride (1:1), Benzeneethanamine, N-ethyl-alpha-methyl-3-(trifluoromethyl)-, hydrochloride (9CI), Phenethylamine, N-ethyl-alpha-methyl-m-(trifluoromethyl)-, hydrochloride (7CI,8CI), (+/-)-Fenfluramine chloride, (+/-)-Fenfluramine hydrochloride, N-Ethyl-alpha-methyl-m-(trifluoromethyl)phenethylamine hydrochloride |
| InChI formel | InChI=1S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16H,3,7H2,1-2H3;1H |
| SMIL | Cl.CCNC(C)Cc1cccc(c1)C(F)(F)F |
| IUPAC navn | N-ethyl-1-[3-(trifluoromethyl)phenyl]propan-2-amine;hydrochloride |
| Vis mere |