missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Fendiline hydrochloride, MedChemExpress
Description
Fendiline hydrochloride is a nonselective calcium channel blocker.
Specifications
Specifications
| Kemisk navn eller materiale | Fendiline hydrochloride |
| Fysisk form | Solid |
| CAS | 13636-18-5 |
| Farve | White |
| Mængde | 100 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C23H26ClN |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Opløselighedsinformation | DMSO : ≥ 30 mg/mL (85.25 mM) ∣H2O : 1.82 mg/mL (5.17 mM; ultrasonic and warming and heat to 60°C) |
| SMIL | CC(NCCC(C1=CC=CC=C1)C2=CC=CC=C2)C3=CC=CC=C3.[H]Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?