Få mere at vide
Fenbuconazole, TRC
Artikelnummer. 30136919
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136919 50 mg
17974415 10 mg
17924465 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136919 Leverandør Toronto Research Chemicals Leverandørnr. TRCF24680050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 114369-43-6
Molekylær formel C19 H17 Cl N4
Molekylvægt (g/mol) 336.82
Synonym α-[2-(4-Chlorophenyl)ethyl]-α-phenyl-1H-1,2,4-triazole-1-propanenitrile, 4-(4-Chlorophenyl)-2-phenyl-2-(1H-1,2,4-triazol-1-ylmethyl)butyronitrile, AG 4452, Fenbuconazole, Fenethanil, Indar, Indar 5, RH 7592
IUPAC navn 4-(4-chlorophenyl)-2-phenyl-2-(1,2,4-triazol-1-ylmethyl)butanenitrile
SMIL Clc1ccc(CCC(Cn2cncn2)(C#N)c3ccccc3)cc1
Tekniske data
Kemisk navn eller materiale Fenbuconazole
Mængde 50 mg
InChI formel InChI=1S/C19H17ClN4/c20-18-8-6-16(7-9-18)10-11-19(12-21,13-24-15-22-14-23-24)17-4-2-1-3-5-17/h1-9,14-15H,10-11,13H2
Formel vægt 336.1142