Få mere at vide
Fenbendazole Sulfone-d3, TRC
Artikelnummer. 30403966
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30403966 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403966 Leverandør Toronto Research Chemicals Leverandørnr. TRCF2467931MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fenbendazole-sulfone D3 (methyl D3)
CAS 1228182-49-7
Molekylær formel C15 D3 H10 N3 O4 S
Mængde 1 mg
Synonym Trideuteriomethyl N-[5-(benzenesulfonyl)-1H-benzimidazol-2-yl]carbamate
InChI formel InChI=1S/C15H13N3O4S/c1-22-15(19)18-14-16-12-8-7-11(9-13(12)17-14)23(20,21)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)/i1D3
SMIL [2H]C([2H])([2H])OC(=O)Nc1nc2ccc(cc2[nH]1)S(=O)(=O)c3ccccc3
IUPAC navn trideuteriomethyl N-[6-(benzenesulfonyl)-1H-benzimidazol-2-yl]carbamate
Molekylvægt (g/mol) 334.36
Formel vægt 334.0815