Få mere at vide
Febuxostat-d7, TRC
Artikelnummer. 30404202
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30404202 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404202 Leverandør Toronto Research Chemicals Leverandørnr. TRCF2290031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Febuxostat-d7
CAS 1285539-74-3
Molekylær formel C16H9D7N2O3S
Mængde 1 mg
Synonym 2-[3-Cyano-4-(2-methylpropoxyphenyl]-4-methyl-5-thiazolecarboxylic Acid-d7, 2-(3-Cyano-4-isobutyloxyphenyl)-4-methyl-5-thiazolecarboxylic Acid-d7, TEI 6720-d7, TMX 67-d7
InChI formel InChI=1S/C16H16N2O3S/c1-9(2)8-21-13-5-4-11(6-12(13)7-17)15-18-10(3)14(22-15)16(19)20/h4-6,9H,8H2,1-3H3,(H,19,20)/i1D3,2D3,9D
SMIL [2H]C(C([2H])([2H])[2H])(C([2H])([2H])[2H])COC1=C(C#N)C=C(C2=NC(C)=C(C(O)=O)S2)C=C1
Molekylvægt (g/mol) 323.42