missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO24 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04452-100UL
Additional Details : Weight : 0.00970kg
Description
FBXO24 Polyclonal specifically detects FBXO24 in Human, Mouse samples. It is validated for Western Blot.Specifications
FBXO24 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
26261 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
DKFZp434I1122, F-box protein 24, F-box protein Fbx24, FBX24F-box only protein 24 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human FBXO24 (NP_277041.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |