missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Fast Skeletal Myosin Heavy Chain 1 Antibody (LM5) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50401JF635
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Fast Skeletal Myosin Heavy Chain 1 Monoclonal antibody specifically detects Fast Skeletal Myosin Heavy Chain 1 in Chicken samples. It is validated for Western Blot
Tekniske data
| Fast Skeletal Myosin Heavy Chain 1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Mouse | |
| Ascites | |
| RUO | |
| Chicken | |
| Ascites |
| Western Blot | |
| LM5 | |
| 50mM Sodium Borate | |
| Purified chicken pectorial muscle | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion