Få mere at vide
Farrerol, TRC
Artikelnummer. 30406962
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30406962 100 mg
30402756 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30406962 Leverandør Toronto Research Chemicals Leverandørnr. TRCF101060100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Farrerol
CAS 95403-16-0
Molekylær formel C17H16O5
Mængde 100 mg
Synonym (±)-4',5,7-Trihydroxy-6,8-dimethyl-flavanone, (±)-Farrerol, rac-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6,8-dimethyl-4H-1-Benzopyran-4-one
InChI formel InChI=1S/C17H16O5/c1-8-15(20)9(2)17-14(16(8)21)12(19)7-13(22-17)10-3-5-11(18)6-4-10/h3-6,13,18,20-21H,7H2,1-2H3
SMIL O=C1C=2C(O)=C(C(O)=C(C2OC(C3=CC=C(O)C=C3)C1)C)C
IUPAC navn 5,7-dihydroxy-2-(4-hydroxyphenyl)-6,8-dimethylchroman-4-one
Molekylvægt (g/mol) 300.31
Formel vægt 300.1