Få mere at vide
Fangchinoline, TRC
Artikelnummer. 30135349
Skift visning
Mængde:
100 mg
250 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135349 250 mg
30136719 2.5 g
30004252 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135349 Leverandør Toronto Research Chemicals Leverandørnr. TRCF102365250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 33889-68-8
Molekylær formel C37H40N2O6
Molekylvægt (g/mol) 608.72
Synonym (4aS,16aR)3,4,4a,5,16a,17,18,19-Octahydro-12,21,26-trimethoxy-4,17-dimethyl-16H-1,24:6,9-dietheno-11,15-metheno-2H-pyrido[2',3':17,18][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolin-22-ol, 6,6',12-Trimethoxy-2,2'-dimethyl-berbaman-7-ol, Thalrugosine, (+)-Thalrugosine, [4aS-(4aR*,16aS*)]-3,4,4a,5,16a,17,18,19-Octahydro-12,21,26-trimethoxy-4,17-dimethyl-16H-1,24:6,9-dietheno-11,15-metheno-2H-pyrido[2',3':17,18][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolin-22-ol, Isofangchinoline, NSC 277171, Thaligine, d-Isofangchinoline
SMIL NCC1OC(C(N)CC1)OC2C(O)C(OC3C(O)C(NC)C(O)(C)CO3)C(N)CC2N
Tekniske data
Kemisk navn eller materiale Fangchinoline
Mængde 250 mg
InChI formel InChI=1S/C37H40N2O6/c1-38-14-12-24-19-31(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-32-18-23(8-11-30(32)41-3)17-29-35-25(13-15-39(29)2)20-34(43-5)36(40)37(35)45-33/h6-11,18-21,28-29,40H,12-17H2,1-5H3/t28-,29-/m0/s1