missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
FAM91A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87432
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FAM91A1 Polyclonal specifically detects FAM91A1 in Human samples. It is validated for Western Blot.
Tekniske data
| FAM91A1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 157769 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| DKFZp666B104, family with sequence similarity 91, member A1, FLJ23790, hypothetical protein LOC157769, skeletal muscle cells re-entry induced | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of human FAM91A1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion