missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM212A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-84909-0.1ML
Additional Details : Weight : 0.00970kg
Description
FAM212A Polyclonal specifically detects FAM212A in Human samples. It is validated for Western Blot.Specifications
FAM212A | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
389119 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
C3orf54, chromosome 3 open reading frame 54, family with sequence similarity 212, member A, INKA1, INKA1 protein, MGC20416 | |
The immunogen is a synthetic peptide directed towards the middle region of Human FAM212A | |
0.1 mL | |
Primary | |
Human | |
Purified |