missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM204A Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-84907-0.1ML
Additional Details : Weight : 0.00970kg
Description
FAM204A Polyclonal specifically detects FAM204A in Mouse samples. It is validated for Western Blot.Specifications
FAM204A | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
63877 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
bA319I23.1, chromosome 10 open reading frame 84, FLJ13188, hypothetical protein LOC63877 | |
The immunogen is a synthetic peptide directed towards the C terminal region of mouse D19ERTD737E | |
0.1 mL | |
Primary | |
Mouse | |
Purified |