Learn More
R&D Systems™ FAM170b Recombinant Protein Antigen
Product Code. 18100970 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1mL
Unit Size:
0.1mL
Product Code. Quantity Unit Size
18100970 0.1mL 0.1mL
1 options
Product Code. 18100970 Supplier R&D Systems™ Supplier No. NBP213985PEP
 Restricted Item
Forventet forsendelse: 21-01-2027
A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human FAM170B. The FAM170b Recombinant Protein Antigen is derived from E. coli. The FAM170b Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP2-13985. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 170370
Species Human
Product Line Novus
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol FAM170B
Label Type Unlabeled
Molecular Weight (g/mol) 24kDa
Product Type FAM170b
Quantity 0.1mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-13985. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen GIREGFSCQIFFEEMLERRRAQGQAHDQQLEEEQSPSDNSECSRPQGEVLSAQQQEKQ
Show More Show Less
For Research Use Only