missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
FAM149B1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06311IR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FAM149B1 Polyclonal specifically detects FAM149B1 in Human samples. It is validated for Western Blot.
Tekniske data
| FAM149B1 | |
| Polyclonal | |
| Western Blot | |
| family with sequence similarity 149, member B1, KIAA0974hypothetical protein LOC317662 | |
| Produced in rabbits immunized with E. coli-derived Human FAM149B1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 317662 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion