missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
FAM102A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-10723-100UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
FAM102A Polyclonal specifically detects FAM102A in Human samples. It is validated for Western Blot.
Tekniske data
| FAM102A | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| bA203J24.7, C230093N12Rik, C9orf132, Early estrogen-induced gene 1 protein, Eeig1, EEIG1chromosome 9 open reading frame 132, family with sequence similarity 102, member A, MGC50853 | |
| The immunogen for Anti-FAM102A antibody is: synthetic peptide directed towards the Middle region of Human F102A. Peptide sequence SLSDLTHRRNTSTSSSASGGLGMTVEGPEGSEREHRPPEKPPRPPRPLHL | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 399665 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion