Learn More
Falcon™ Pipet Controller Replacement Parts
GREENER_CHOICE
Assortment of replacement parts for pipet controllers.
Brand:  Falcon™ 357489
236.20 DKK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 15359860
  • 304.51 DKK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
Estimated Shipment: 06-11-2025
Description
For use with Falcon pipet controller.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal Power Supply
Pipet controller
In: 100/250 V, Out: 9 V
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Falcon™ Pipet Controller Replacement Parts > Universal Power Supply; In: 100-250V, Out: 9V