Få mere at vide
FAK Inhibitor 14, TRC
Artikelnummer. 30179172
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179172 1 g
30179429 500 mg
30179501 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179172 Leverandør Toronto Research Chemicals Leverandørnr. TRCF1010731G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4506-66-5
Molekylær formel C6H10N4 . 4(HCl)
Molekylvægt (g/mol) 284
Synonym 1,2,4,5-Benzenetetramine Tetrahydrochloride, 1,2,4,5-Benzenetetramine, tetrahydrochloride (6CI,7CI,8CI,9CI), NSC 667249, Y 15
SMIL Cl.Cl.Cl.Cl.Nc1cc(N)c(N)cc1N
Tekniske data
Kemisk navn eller materiale FAK Inhibitor 14
Mængde 1 g
InChI formel InChI=1S/C6H10N4.4ClH/c7-3-1-4(8)6(10)2-5(3)9;;;;/h1-2H,7-10H2;4*1H