Learn More
Memmert™ Factory Calibration (3 Temperatures) for UN160
Calibration at Memmert factory for three temperature value.
Brand:  Memmert™ UN160D00128
Product Code. 17569865
  • 2655.00 DKK / Each
Description
  • For three temperature values: 100°C, 160°C, and 220°C
Specifications
Specifications
UN160 Oven
Measuring point chamber center
Certificate for 3 temperatures
Standard Works Calibration Certificate
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Factory Calibration (3 Temperatures) for UN160 > For Use With: UN160