missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550407
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total FABP6 protein.
FABP6 Polyclonal antibody specifically detects FABP6 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FABP6 | |
Polyclonal | |
Unconjugated | |
FABP6 | |
FABP6; fatty acid binding protein 6; fatty acid binding protein 6, ileal; fatty acid binding protein 6, ileal (gastrotropin); fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ileal bile acid binding protein; ileal bile acid-binding protein; ileal lipid-binding protein; Illbp; illeal lipid-binding protein; intestinal 15 kDa protein; Intestinal bile acid-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16204, 2172 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P51161, P51162 | |
FABP6 | |
Full length fusion protein of Human FABP6 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |